Learn More
3M™ Specialty Disposable Unvalved Respirator
Provides lightweight, effective, comfortable and hygienic respiratory FFP1 protection against dust, mist and nuisance-level acid gases. 3M™ Specialty Disposable Unvalved FFP1 Respirators have a robust shell and cup design which conforms well to most face shapes and sizes and is collapse resistant.
Brand:  3M™ 7000034737
14.97 EUR valid until 2024-03-30
Use promo code "21636" to get your promotional price.
Additional Details : Weight : 0.34380kg
Product Code. 12362399
  • 148.10€ / Pack of 20
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
Description
  • Activated carbon layer
  • Convex shape, nose clip and twin-strap design
  • Maximum usage level: up to 4 x TLV for particulates; below TLV for acid gases