missing translation for 'onlineSavingsMsg'
Learn More

Serva Electrophoresis™ Spectra/Por™ 3 Dialysis Tubing

Product Code. 11762477
Encompass_Preferred
Change view
Click to view available options
MWCO:
3.5 kDa
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO unitSize
11762477 3.5 kDa Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11762477 Supplier Serva Electrophoresis™ Supplier No. 4418501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Dialysis Tubing
MWCO 3.5 kDa
Material Cellulose
Diameter (Metric) 34 mm
Length (Metric) 15 m
Capacity (Metric) 9.3 mL/cm
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.