Learn More
Serva Electrophoresis™ Spectra/Por™ 6 Dialysis Tubing
Specifications
Specifications
| Product Type | Dialysis Tubing |
| MWCO | 8 kDa |
| Diameter (Metric) | 25.5 mm |
| Length (Metric) | 10 m |
| Capacity (Metric) | 5.1 mL/cm |
| Quantity | 1 Each |