missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 7 8000 D MWCO Standard RC Pre-Treated Dialysis Kits

With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds

Brand:  Spectrum™ 132 114T

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15380692

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Includes:
  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks

Description

Description

  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application

Specifications

Specifications

8 kDa
15 mm
1.8 mL/cm vol./length
1 m Tubing/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions