Learn More
Spectrum™ Spectra/Gel™ Absorbent
Product Code. 10639975
Click to view available options
:
500g
Product Code. 10639975
Brand: Spectrum™ 292600
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Makes the concentration of protein solutions quick and efficient
  • Material: Polyacrylate-polyalcohol
  • Expands when it absorbs water
  • Can be used with dialysis membranes with MWCOs from 0.1 to 300 kD
  • Easy to remove
Specifications
Product Type Absorbent
MWCO 0.1 to 300 kDa
Material Polyacrylate-Polyalcohol
Color White
Quantity Each
For Use With (Application) For use with any MWCO dialysis membrane, high water absorbent capacity, convenient for sample concentration with Float-A-Lyzer™ G2
Length of time needed for concentration depends on the final volume, MWCO, surface-to-volume ratio of membrane tubing and amount of absorbent.