Learn More
Cole-Parmer™ Specwell™ M820-S Microscope/Telescope with Built-in Reticle
Shop All Cole-Parmer Products
Click to view available options
Magnification Power:
8x/25x
Unit Size:
Each
Description
- Scope are constructed of durable aluminum and plastic
- Lenses come with covers to protect against scratches
- Eyepiece has a soft edge for comfort and safety
- Includes a neck strap, a micro lens stand, hard carrying case and carrying strap
- Ideal for macroscopic quality control and inspection procedures, this model includes a straight line reticle
- For microscopic viewing, screw the micro lens and stand onto the monocular
- For distant telescopic viewing, unscrew the micro lens and stand and use the monocular as a telescope (these scopes are not intended for close range telescopic viewing)
- Focus is adjustable in both microscope and telescope modes
Specifications
Specifications
| Type | Microscope/Telescope with Built-in Reticle |
| Diameter (English) | 1.06 in. |
| Diameter (Metric) | 2.7 mm |
| Field of View | 9 mm |
| Height (English) | 7.08 in. |
| Height (Metric) | 18 cm |
| Includes | Neck strap, a micro lens stand, hard carrying case and carrying strap |
| Magnification Power | 8x/25x |
| Model | Specwell™ M820-S |