missing translation for 'onlineSavingsMsg'
Learn More

SPEXQuE™ GC/MS Internal Standard Mix, SPEX CertiPrep™

Sensible solutions for pesticide extraction

Brand:  SPEX Certiprep QUECR-MS-IS

Additional Details : Weight : 0.00100kg Transport : UN number : 1648 Chem class : 3 Pack group : II


Product Code. 16319997

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

  • Solvent: Acetonitrile
  • Volume: 5mL

Specifications

Specifications

50 µg/mL
5 mL
GC/MS Internal Standard Mix
Acetonitrile
Pesticide Extraction
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GC/MS Internal Standard Mix

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : acetonitrile

SDS
Documents

Documents

Certificates
Promotions

Promotions