missing translation for 'onlineSavingsMsg'
Learn More

SPEX™ SamplePrep 6775 Freezer/Mill™ Accessories: Vial Set

For use with Spex SamplePrep Freezer/Mill Cryogenic Grinder

Brand:  SPEX™ SamplePrep 6754

Additional Details : Weight : 5.00000kg

 View more versions of this product

Product Code. 15136423

Quantity Price
1 945.08€ / 1 unit
4 817.68€ / 1 unit
Estimated Shipment
28-05-2021
Add to basket


Description

Description

Vial Set 6751

  • Sample capacity: 0.5 to 4.0 mL
  • Makes one complete vial plus three spare center sections
  • Includes stainless-steel impactor, two end plugs, and four polycarbonate center cylinders (6751C4)
  • Individual components available

Specifications

Specifications

Ideal to Remove The End Plugs From The Center Cylinders of Small Grinding Vials
0.5 to 4 mL
Extractor and Vial Opener Set
0.01 to 0.13 oz.
Spex SamplePrep Freezer/Mill Cryogenic Grinder
Videos
SDS
Documents