Learn More
Cole-Parmer™ Replacement Sample Extraction Tool for Microvial Grinding Set
Product Code. 12520196 Shop All SPEX Certiprep Products
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
Product Code. 12520196
Brand: Cole-Parmer™ 6753C
This item requires a price update prior to being ordered.
Request a Price for this item here.
Sample Extraction Tool for removing ground samples from microvials which are used in the microvial set.
  • Insert the tool into an individual vial, then push it through to clear sample from the tube.
Specifications
Product Type Replacement Extraction Tool
For Use With (Equipment) 6757 Grinding Microvial Set (61043-06)
Packaging Quantity 1 Ea.