Learn More
Cole-Parmer™ Vial Set for High-Energy Mixer/Mill™ Grinders
Shop All SPEX Certiprep Products
Click to view available options
Material:
Agate
Alumina Ceramic
Hardened Steel
Stainless Steel
Tungsten Carbide-lined
Zirconia
Diameter (Metric):
1.27 cm
1.9 cm
5.7 cm
5.71 cm
5.72 cm
6.03 cm
6.35 cm
Height (Metric):
2.54 cm
4.76 cm
5.08 cm
6.83 cm
6.98 cm
7.62 cm
Unit Size:
1 set
Each
Description
- Crush, grind, and mill various materials.
- Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinders with a wide range of vial sets suitable for your application.
Specifications
Specifications
| Product Type | Grinding Vial Set |
| Diameter (English) | 2.5 in. |
| Diameter (Metric) | 6.35 cm |
| For Use With (Equipment) | 8000 D, 8000 M Mixer/Mills |
| Height (English) | 2.69 in. |
| Height (Metric) | 6.83 cm |
| Includes | Two 1/2 in. Grinding Balls |
| Material | Zirconia |