Learn More
Cole-Parmer™ Grinding Balls for High-Energy Mixer/Mill™ Grinding Vial Sets
Product Code. 12538706 Shop All SPEX Certiprep Products
Click to view available options
Material:
Hardened Steel
Stainless Steel
Unit Size:
Each
Pack of 6
Product Code. 12538706
Brand: Cole-Parmer™ 8007B
This item requires a price update prior to being ordered.
Request a Price for this item here.
Crush, grind, and mill various materials with these Grinding Balls for High-Energy Mixer/Mill™ Grinding Vial Sets.
  • Select from 10 different grinding ball materials to suit your application.
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.
Specifications
Product Type Grinding Balls
Diameter (English) 0.5 in., 0.25 in.
Diameter (Metric) 1.27 cm, 0.63 cm
For Use With (Equipment) Vial Kit of 8000 D and 8000 M Mixer or Mill
Includes Two 0.5 in. and four 0.25 in. hardened 440 C Stainless Steel balls
Material Stainless Steel