Learn More
Huenersdorf™ Square Lid
Brand: Huenersdorf™ 945100
Additional Details : Weight : 0.18000kg
Specifications
Lid, Square | |
45 mm | |
1 Ea. |
Natural | |
Lid | |
280 mm |
Brand: Huenersdorf™ 945100
Additional Details : Weight : 0.18000kg
Lid, Square | |
45 mm | |
1 Ea. |
Natural | |
Lid | |
280 mm |