Learn More
Cole-Parmer™ Rubbermaid™ Square Scoop
Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
2 L
Unit Size:
Each
Description
Scoop holds 2 quarts. Stands on end or hangs from a hook.
Specifications
Specifications
| Capacity (Metric) | 2 L |
| Handle Type | Molded |
| Length (Metric) | 210 mm |