missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Squarunkin A Hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Squarunkin A hydrochloride is a potent inhibitor of UNC119 chaperone protein-cargo interaction (IC50 = 10 nM for inhibition of UNC119A - myristoylated Src N-terminal peptide interaction). Disrupts activation of Src kinase in vitro.
Specifications
Specifications
| Chemical Name or Material | Ethyl 4-((3,4-dioxo-2-((2-(4-(3-(trifluoromethyl)phenyl)piperazin-1-yl)ethyl)amino)cyclobut-1-en-1-yl)amino)piperidine-1-carboxylate hydrochloride |
| Quantity | 10 mg |
| Molecular Formula | C25H32F3N5O4.HCl |
| Formula Weight | 560.01 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction