Learn More
Fisherbrand™ Squat Form Beakers
Product Code. 11972203 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
800 mL
1000 mL
2000 mL
3000 mL
5000 mL
10 L
Unit Size:
Each
Pack of 10
Product Code. 11972203
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Constructed of borosilicate glass. Fisherbrand™ Squat Form Beakers are graduated and come in a large variety of capacities.
  • Borosilicate glass
  • Squat form with spout and graduations
  • ISO 3819 DIN 12331
TRUSTED_SUSTAINABILITY
Specifications
Color Translucent
Diameter (Metric) Outer 170 mm
Capacity (Metric) 5000 mL
Material Borosilicate Glass
Finish Matte
Height (Metric) 270 mm
Includes Spout
Quantity 1 Ea.
Type Beaker
Graduated Yes
Certifications/Compliance ISO 3819, DIN 12331
Scale White Graduated Scale
Style Low Form