missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 57227 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A high affinity, selective 5-HT3 receptor agonist, active in vivo and CNS penetrant.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 248.15 |
| Purity | >99% |
| Molecular Weight (g/mol) | 248.151 |
| Chemical Name or Material | SR 57227 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction