Learn More
Gilac™ Stackable Perforated Crate with Handles
Click to view available options
:
Gilac™ Stackable Perforated Crate
Description
- Its compact size of 500 x 400 mm, perforated walls and base, and handles for a secure grip make this crate an essential daily ally
- A stackable crate of professional quality
Specifications
Specifications
| Closure Type | None |
| Color | Gray |
| Material | HDPE |
| Shape | Rectangular |
| Product Type | Stackable Perforated Crate |
| Dimensions (L x W x H) | 500 x 400 x 330 mm |
| Length (Metric) | 500 mm |
| Width (Metric) | 400 mm |
| Height (Metric) | 330 mm |