Learn More
Cytiva Stainless Steel 316 T-Junction
Product Code. 11310232 Shop All Cytiva Products
Click to view available options
Diameter (Metric) Inner:
6 mm
Unit Size:
Each
Product Code. 11310232
Brand: Cytiva 18110429
Only null left
Specifications
Connection Type 2 x 25 mm TC, 1 x 51 mm TC
Material Stainless Steel
Quantity 1 Each
Diameter (Metric) Inner 6 mm