Learn More
Zwiebel™ Stainless Steel Antimagnetic Weights Handling Clamp
Product Code. 11758214
Click to view available options
Length (Metric):
10 cm
15 cm
Unit Size:
Each
Product Code. 11758214
Brand: Zwiebel™ PBI2812E
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use these clamps for the handling of Zwiebel™ weights.
  • Stainless steel handling clamp
  • The pieces in contact with the weight are protected rubber
  • Available in a variety of sizes
Specifications
Material Anti-magnetic Stainless Steel
Length (Metric) 10 cm
Product Type Handling Clamp
For Use With (Application) For handling weights of 1mg to 50g
Quantity 1Each
Weight (Metric) 13g