Learn More
Bochem™ Stainless Steel Beakers
Product Code. 10603511
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Includes:
Rim
Rim and Spout
Rim, Handle
Rim, Spout
Rim, Spout, Handle
Unit Size:
Each
Product Code. 10603511
Brand: Bochem™ 8503
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Highly resistant and durable stainless steel beaker. Bochem™ Stainless Steel Beakers include rims, spouts, and handles in various model combinations.
  • 18/10 stainless steel construction.
  • Resistant and durable.
  • Contact bottom for heating plates and magnetic stirrers.
Specifications
Capacity (Metric) 1000 mL
Material Stainless Steel (18/10)
Includes Rim
Type Beaker