Learn More
Sampling Systems™ Stainless Steel Bottle
Store liquids and powders securely with this range of high quality Stainless Steel Containers. Sampling Systems™ Stainless Steel Bottle is ideal for storing and transporting powders and liquids.
291.00€ - 299.00€
Specifications
Material Stainless Steel (316L)
Closure Type Tri Ferrule
View More Specs
Products 2
Product Code Brand Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
15819721
View Documents Promotion Details
Sampling Systems™
A836-1000
1000 mL 102 mm 178 mm
299.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 10-07-2024
to see stock.
15829721
View Documents Promotion Details
Sampling Systems™
A836-500
500 mL 83 mm 148 mm
291.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Description
Description
  • Multi-purpose containers
  • Robust construction
  • Crevice free interior
  • Standard lid (tri-ferrule style BS 4825-3)
  • All welds ground and polished
  • Complete with lid, silicone gasket (FDA Compliant) and clampband
  • 316L stainless steel body and lid
Specifications
Specifications
Stainless Steel (316L)
Tri Ferrule