Learn More
RETSCH Stainless Steel Bottom Sieve For SM 200 Cutting Mills
Shop All Retsch Products
Click to view available options
Pore Size:
1 mm
1.5 mm
2 mm
250 μm
4 mm
500 μm
6 mm
750 μm
8 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 500 μm |
| Type | Bottom Sieve |
| Material | Stainless Steel |
| For Use With (Equipment) | SM 200 Cutting Mill |