Learn More
Sampling Systems™ Stainless Steel Cable
Product Code. 15809351 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
10 m
25 m
50 m
Unit Size:
10m
25m
50m
Product Code. 15809351
Brand: Sampling Systems™ 5070D10
Only null left
Add to basket
Add to basket
Sampling Systems™ Stainless Steel Cable is designed to be used in conjuction with a Disposable Bailer with or without a weighted end.
  • Cable for Disposable Bailer
  • PTFE coated Stainless Steel
  • Avalable in a variety of lengths
Specifications
Product Type Cable
Material Stainless Steel
Finish PTFE
For Use With (Equipment) Disposable Bailer
Length (Metric) 10 m