Learn More
Sampling Systems™ Stainless Steel Cable
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
10 m
25 m
50 m
Unit Size:
10m
25m
50m
Description
- Cable for Disposable Bailer
- PTFE coated Stainless Steel
- Avalable in a variety of lengths
Specifications
Specifications
| Product Type | Cable |
| Material | Stainless Steel |
| Finish | PTFE |
| For Use With (Equipment) | Disposable Bailer |
| Length (Metric) | 50 m |