Learn More
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire
Product Code. p-7188337 Shop All Ohaus Products
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
Each
Product Code. 16662292
Brand: Ohaus™ 30517704
Only null left
Add to basket
Add to basket
Adjusted to Meet or Exceed OIML E1 Tolerances
  • OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance No Certificate
Material Stainless Steel (316)
Mass 1 mg
Tolerance 0.003 mg
Includes Protective Case