Learn More
Amsta Rostfritt Stainless Steel Containers
Product Code. 11761786
Click to view available options
Volume (Metric):
1 L
2 L
3 L
5 L
6 L
10 L
50 L
Includes:
3 clamps, without lid
With Lid
Without Lid
missing translation for 'unitSize'
Each
Product Code. 11761786
missing translation for 'mfr': Amsta Rostfritt 5060321
Only null left
Add to basket
Add to basket
With round handle in highly polished finish.
Specifications
Product Type Container with Lid
Material Stainless Steel
Dimensions (L x W) 130 x 170 mm
Includes With Lid
Volume (Metric) 2 L