Learn More
De Buyer™ Stainless Steel Dishes
Product Code. 11980615
Click to view available options
Height (Metric):
4 cm
Unit Size:
Each
Product Code. 11980615
Brand: De Buyer™ 328027
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Bottom Shape Flat
Capacity (Metric) 1.9 L
Material Stainless Steel
Shape Rectangular
Type Dish
Length (Metric) 27 cm
Width (Metric) 22 cm
Height (Metric) 4 cm