Learn More
Agencinox™ Stainless Steel Double Spatula
Product Code. 11706715
Click to view available options
Length (Metric):
195 mm
230 mm
Unit Size:
Pack of 5
Product Code. 11706715
Brand: Agencinox™ 1869
Only null left
Add to basket
Edge
Add to basket
Transfer your samples without contamination, sticking and the assurance of complete release. Agencinox™ Stainless Steel Double Spatulas are chemically inert and manufactured with smooth, flat surfaces.
  • Stainless steel double spatula
  • Available with blades and handles of varying width and lengths
  • Ideal for most laboratory applications
Specifications
Quantity 5 Pk.
Certifications/Compliance CE, ISO
Product Type Double Ended Spatula
Material Stainless Steel
Length (Metric) 195 mm