missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Ferritic, AISI 409, S40900, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Ferritic, AISI 409, S40900, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Zirconium/Lead/Boron/Sulfur/Antimony/Arsenic/Tungsten/Niobium/Tin/Carbon/Nitrogen/Zinc/Molybdenum/Aluminium/Cobalt/Phosphorus/Copper/Vanadium/Nickel/Titanium/Manganese/Silicon/Chromium/Iron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium -100 ppm/Zirconium -21 ppm/Lead -10 ppm/Boron -3 ppm/Sulfur 0.0007 %/Antimony 0.0011 %/Arsenic 0.0026 %/Tungsten 0.0036 %/Niobium 0.0041 %/Tin 0.0053 %/Carbon 0.01 %/Nitrogen 0.01 %/Zinc 0.01 %/Molybdenum 0.011 %/Aluminium 0.017 %/Cobalt 0.019 %/Phosphorus 0.021 %/Copper 0.0655 %/Vanadium 0.075 %/Nickel 0.11 %/Titanium 0.141 %/Manganese 0.387 %/Silicon 0.52 %/Chromium 11.28 %/Iron (87.4) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction