Learn More
Bochem™ Stainless Steel Forceps
Click to view available options
Length (Metric):
125 mm
Unit Size:
Each
Description
- For use with filter paper
Specifications
Specifications
| No. per Pack | 1 Pack |
| Material | Stainless Steel |
| Length (Metric) | 125 mm |
| Product Type | Forceps |
| For Use With (Application) | For filter paper |
| Width (Metric) Tip | 12 mm |
| Width (Metric) | 12 mm |
| Type | Forceps |