Learn More
Stainless Steel gauze, 100 mesh woven from 0.11mm (0.0045in) dia wire, Type 316, Thermo Scientific Chemicals
29.60€ - 89.50€
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15494065
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046703.FL
75 x 75 mm
29.60€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
15404075
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046703.GT
150 x 150 mm
49.70€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
15414075
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046703.HB
300 x 300 mm
89.50€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
Stainless Steel gauze, type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.