missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel gauze, 100 mesh woven from 0.11mm (0.0045in) dia wire, Type 316, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 046703.GT

Additional Details : Weight : 0.01000kg


 View more versions of this product

Product Code. 15404075

  • 49.70€ / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Stainless Steel gauze, type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stainless Steel gauze, Type 316
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
150 x 150 mm
100 Mesh
Wire Cloth
100 mesh woven from 0.11mm (0.0045 in.) dia. wire
MFCD00148499
Insoluble in water.
Odorless
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Stainless Steel gauze, Type 316

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Iron, Chromium, Nickel

SDS Documents

Documents

Certificates
Promotions

Promotions

RUO – Research Use Only