Learn More
Trajan Scientific and Medical™ Stainless-Steel GC Tubing
Product Code. p-7114376
Click to view available options
Overall Length (Metric):
10 m
2 m
25 m
5 m
Diameter (Metric) Inner:
0.25 mm
0.50 mm
0.8 mm
2.2 mm
Unit Size:
2m
Each
Product Code. 15258723
Brand: Trajan Scientific and Medical™ 0624173
Only null left
Add to basket
Add to basket
Features a smooth, clean bore for use in HPLC systems.
  • Smooth and clean bore
  • Supplied in coils
  • Supplied cleaned and passivated
Specifications
Type Tubing
Includes Smooth and clean bore
Diameter (Metric) Inner 2.2 mm
Diameter (Metric) Outer 3.1 mm
Overall Length (Metric) 2 m