missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Greek Ascoloy, AISI 418, S41800, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, Greek Ascoloy, AISI 418, S41800, Ø30x3mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Aluminium/Tin/Oxygen/Niobium/Phosphorus/Nitrogen/Cobalt/Copper/Vanadium/Molybdenum/Carbon/Manganese/Silicon/Nickel/Tungsten/Chromium |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.003 %/Aluminium 0.004 %/Tin 0.004 %/Oxygen 0.0068 %/Niobium 0.01 %/Phosphorus 0.018 %/Nitrogen 0.0222 %/Cobalt 0.031 %/Copper 0.06 %/Vanadium 0.086 %/Molybdenum 0.12 %/Carbon 0.18 %/Manganese 0.3 %/Silicon 0.35 %/Nickel 1.93 %/Tungsten 2.59 %/Chromium 12.15 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction