Learn More
Alpaco™ Stainless Steel Clamps with Vertical Screws
Product Code. 11949717
Click to view available options
Size (Metric) Grip:
12 to 17 mm
14 to 19 mm
16 to 21 mm
17 to 22 mm
20 to 25 mm
22 to 27 mm
25 to 30 mm
28 to 33 mm
30 to 35 mm
36 to 41 mm
5 to 10 mm
7 to 12 mm
9 to 14 mm
Unit Size:
Pack of 10
Product Code. 11949717
Brand: Alpaco™ INH 14/101615
Only null left
Add to basket
Add to basket
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Stainless Steel Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
  • Reusable
  • Resists pressure
  • No lubricant required for assembly
  • Smooth clamping to avoid tubing damage
Specifications
Quantity 10/Pk.
Type Clamp
Material Stainless Steel
Size (Metric) Grip 9 to 14 mm