Learn More
Bochem™ Stainless Steel Lid
42.90€ - 158.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10005250
|
Bochem™
8332 |
For 10 L Graduated Bucket |
42.90€
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
10634121
|
Bochem™
8392 |
For 10 L Graduated measuring jug |
137.00€
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
10107440
|
Bochem™
8334 |
For 15 L Graduated Bucket |
49.70€
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10694301
|
Bochem™
8393 |
For 15 L Graduated measuring jug |
158.00€
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
Specifications
Lid | |
Stainless Steel (18/10) |