Learn More
Linker Industrietechnik™ Stainless Steel Lid with Sprinkler Mechanism for 200mm dia.Sieves
Cover sieves to avoid loss of sample during separation process. This stainless steel lid includes a sprinkler mechanism, fits 200mm diameter sieves, and is recommended for sieves with 25mm height.
Brand:  Linker Industrietechnik™ 552.025.200-910
Additional Details : Weight : 0.38000kg
Product Code. 11754429
  • 289.38€ / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
Stainless steel lid can be used with 200mm diameter test sieves that have an internal height of 25mm. Sprinkler mechanism includes 3/4 in. hose connector.
Specifications
Specifications
For 200 dia. x 25mm internal height sieves
200 mm
Lid with Sprinkler Mechanism