Learn More
Cole-Parmer™ Stainless Steel Luer Fittings
Exceptionally durable Luer fittings suitable for harsh chemical environments. Cole-Parmer™ Stainless Steel Luer Fittings are easy to disassemble for cleaning.
Brand: Cole-Parmer™ 41507-21
Additional Details : Weight : 0.00453kg
Description
Exceptionally durable Luer fittings suitable for harsh chemical environments. Cole-Parmer™ Stainless Steel Luer Fittings are easy to disassemble for cleaning.
- Exceptional durability
- Easy to disassemble for cleaning
- Ideal for harsh chemical environments
- Autoclavable
Specifications
Luer Fitting | |
Stainless Steel | |
1 Pack |
Autoclavable | |
Harsh chemical environments | |
Female Luer x Male Luer Lock |