Learn More
Hamilton™ Custom Kel-F Hub Needles
Click to view available options
Needle Gauge:
10 G
14 G
16 G
17 G
20 G
22 G
22s G
25 G
28 G
30 G
Needle Point Style:
2, 3, or 4
5 or AS
Custom
Unit Size:
Pack of 6
Description
- Available in various gauges
- Provided in packs of 6
- Custom point style
- Autoclavable
- Gas sterilizable
- For Kel-F hubs
- Custom length
- Stainless steel Luer tip
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Needle Gauge | 28 G |
| Diameter (Metric) Inner | 0.18 mm |
| Diameter (Metric) Outer | 0.36 mm |
| Product Type | Luer Lock Kel-F Hub Needle |
| Quantity | 6/Pk. |
| Needle Point Style | 2, 3, or 4 |
| For Use With (Equipment) | LT and TLL Syringes and Connectors |
| Connection Type | Standard Tapered |