Learn More
Bel-Art™ Scienceware™ Stainless Steel Micro Spatula
Shop All SP Scienceware Products
Click to view available options
Length (Metric):
5 cm
Unit Size:
Pack of 2
Specifications
Specifications
| Quantity | 2 Pk. |
| Coating | Fluo-Kem Teflon FEP |
| Certifications/Compliance | Complies with FDA regulations |
| Product Type | Micro Spatula |
| Width (Metric) | 0.79 cm |
| Diameter (Metric) Outer | 4mm |
| Dimensions (L x W) | 5 x 0.79 cm |
| Material | Stainless Steel |
| Length (Metric) | 5 cm |