missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Stainless Steel OIML Class E2 Service Weight Sets Without DAkkS Calibration Certificate

E2 Service weight set, ranging from 1mg to 10kg

Brand:  Sartorius™ YSS5128-6528-00

Additional Details : Weight : 0.60000kg

Product Code. 10707812

  • 3830.00€ / 1 set
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

  • E2 weight set
  • From 1mg to 10kg
  • Special steel, non-megnetizable
  • Highly polished
  • In wooden case
Specifications

Specifications

Highly polished E2 weight set in wooden box
Special Steel, Non-magnetizable
1 mg to 10 kg
Service weigt / calibration
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions