Learn More
Bel-Art™ Stainless Steel Pharma Scoops
Product Code. 16874146 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
10 mL
50 mL
100 mL
500 mL
750 mL
1000 mL
1250 mL
1700 mL
2500 mL
Unit Size:
Each
Product Code. 16874146
Brand: Bel-Art™ F367381250
Only null left
Add to basket
Add to basket
Seamless design for peace of mind
  • Reusable, high-quality scoops can be used for powder and granule handling in pharmaceutical and food handling settings
  • Pharmaceutical grade 316 Stainless Steel
  • Easy to clean seamless design to help prevent cross-contamination
  • High salt and acid resistance, ideal for food sampling
  • GMP compliant
  • Autoclavable
Specifications
Autoclavable Autoclavable
Capacity (English) 44 oz.
Capacity (Metric) 1250 mL
Product Type Scoop
Material Stainless Steel