Learn More
Sampling Systems™ Stainless Steel PharmaBottles™
Product Code. 15849721 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Diameter (Metric):
83 mm
102 mm
Height (Metric):
154 mm
178 mm
Unit Size:
Each
Product Code. 15849721
Brand: Sampling Systems™ A846500
Sampling Systems™ Stainless Steel PharmaBottles™ are ideal for use with powders and liquids.
  • Multi-purpose containers
  • 316L Stainless Steel body and lid
  • Robust construction
  • Crevice free interior
  • All welds ground and polished
  • Complete with screw on lid
  • Silicone (FDA acceptable) gasket in lid
Specifications
Material Stainless Steel (316L)
Capacity (Metric) 500 mL
Closure Type Threaded Cap
Diameter (Metric) 83 mm
Height (Metric) 154 mm