Learn More
Schneider Gerd™ Stainless Steel Pharmacists Spatula
Use this spatula for a variety of applications. Schneider Gerd™ Stainless Steel Pharmacists Spatulas are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 610
Additional Details : Weight : 0.02700kg
 View more versions of this product
Product Code. 10103004
  • 15.75€ / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Spatulas are ideal for working with powders, stirring and mixing
  • Perfect for miniscule amounts of chemicals
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
1 Ea.
Stainless Steel
Stainless Steel
185 mm
Pharmacists Spatula
Wood
Wood