missing translation for 'onlineSavingsMsg'
Learn More

Smeg™ Stainless Steel Pipette Washing Trolley

Designed for use with Smeg™ pipette washing devices. Smeg Stainless Steel Pipette Washing Trolleys can wash and dry up to 100 pipettes with a capacity of 1 to 20mL.

Brand:  Smeg™ LPT100

Additional Details : Weight : 0.80000kg

Product Code. 11935624

  • 2400.00€ / Each
Estimated Shipment: 13-05-2024
to see stock.
Add to basket

Description

Description

  • Stainless steel pipette washing trolley with drying system connection
  • For washing and drying of 100 pipettes with a capacity of 1 to 20mL and a height up to 450mm
  • The pipette washing disc is fitted in a trolley to allow fast loading and unloading access
Specifications

Specifications

Pipette Washing Trolley
For washing and drying of 100 pipettes with a capacity of 1 to 20mL and a height up to 450mm
Smeg glassware washers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions