Learn More
RUBIS High Precision Tweezers
Click to view available options
Length (Metric):
90 mm
110 mm
115 mm
120 mm
135 mm
Tip Style:
Conical, Ultra Fine
Fine
Pointed, Thick
Pointed, Ultra Fine
Round, Flat
Design:
Straight
Straight, Self Locking
Straight, Self-Locking
Straight, Strong, Sturdy
Unit Size:
Each
Description
- High Precision Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Perfect alignment
- Very effective
- Reliable
- Excellent chemical resistance
Specifications
Specifications
| Tip Style | Round, Flat |
| Design | Straight, Self-Locking |
| Length (English) | 4.72 in. |
| Material | Anti-acid, Anti-magnetic Stainless Steel |
| Length (Metric) | 120 mm |
| Product Type | High Precision Tweezers |
| Quantity (No. per Case) | Each |
| Type | Tweezers, Lock |