Learn More
RUBIS High Precision Tweezers
Product Code. 11773266
Encompass_Preferred
Click to view available options
Length (Metric):
90 mm
110 mm
115 mm
120 mm
135 mm
Tip Style:
Conical, Ultra Fine
Fine
Pointed, Thick
Pointed, Ultra Fine
Round, Flat
Design:
Straight
Straight, Self Locking
Straight, Self-Locking
Straight, Strong, Sturdy
Unit Size:
Each
Product Code. 11773266
Brand: RUBIS 0C9
Only null left
Add to basket
Add to basket
High Precision Tweezers - Antimagnetic/Anti-acid SS
  • High Precision Tweezers
  • 100% Made in Switzerland
  • Anti-acid/Antimagnetic Stainless Steel
  • Perfect alignment
  • Very effective
  • Reliable
  • Excellent chemical resistance
Specifications
Tip Style Pointed, Ultra Fine
Design Straight
Length (English) 3.54 in.
Material Anti-acid, Anti-magnetic Stainless Steel
Length (Metric) 90 mm
Product Type High Precision Tweezers
Quantity (No. per Case) Each