Learn More
Cole-Parmer™ Stainless Steel Pressure Filtration Holder
Brand: Cole-Parmer™ 02926-32
Additional Details : Weight : 1.00000kg
Specifications
Autoclavable | |
Stainless Steel | |
200 mL | |
69.85 mm |
Pressure Filtration | |
1 Each | |
361.95 |