Learn More
RUBIS High Precision Tweezers - SwissTweeze
High Precision Tweezers - Antimagnetic/Anti-acid SS, Perforated Handle
21.03€ - 29.95€
Specifications
Material | Anti-acid, Anti-magnetic Stainless Steel |
---|---|
Product Type | High Precision Tweezers |
Quantity (No. per Case) | Each |
Product Code | Brand | Design | Tip Style | Length (Metric) | Length (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Design | Tip Style | Length (Metric) | Length (English) | Price | Quantity & Availability | ||
11789564
|
RUBIS
5AG |
Angled | Pointed, Ultra Fine | 115 mm | 4.52 in. |
27.54€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
11779564
|
RUBIS
4G |
Straight | Needle Pointed, Ultra Fine | 110 mm | 4.32 in. |
24.13€
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
11799564
|
RUBIS
5ARG |
Angled, Bent | Pointed, Ultra Fine | 115 mm | 4.52 in. |
29.95€
Each |
In Stock
Log in to see stock. |
||
11749564
|
RUBIS
1G |
Straight | Pointed, Ultra Fine | 120 mm | 4.72 in. |
21.03€
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
||
11759564
|
RUBIS
2G |
Straight | Pointed, Fine | 115 mm | 4.52 in. |
22.63€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||
11709574
|
RUBIS
7G |
Curved | Pointed, Ultra Fine | 115 mm | 4.52 in. |
28.74€
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11769564
|
RUBIS
2AG |
Straight | Round, Flat, Fine | 110 mm | 4.52 in. |
22.24€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||
Description
- High Precision Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Perfect alignment
- Very effective
- Reliable
- Excellent chemical resistance
- Swiss Tweeze series
- Perforated handle for enhanced grip
Specifications
Anti-acid, Anti-magnetic Stainless Steel | |
High Precision Tweezers | |
Each |