Learn More
Bochem™ Stainless Steel Rods
Product Code. 10484091
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
1500 mm
Length (Metric) Rod:
500 mm
600 mm
750 mm
1000 mm
1500 mm
Unit Size:
Each
Product Code. 10484091
Brand: Bochem™ 5122
Only null left
Add to basket
Add to basket
Use Bochem™ Stainless Steel Rods for strength for your lab operations.
  • Stainless Steel
  • Sturdy
Specifications
Material Stainless Steel
Length (Metric) 750 mm
Length (Metric) Rod 750 mm
Diameter (Metric) Rod 12 dia.
For Use With (Application) Stand Bases